missing translation for 'onlineSavingsMsg'
Learn More

Sodium glycocholate hydrate, 98%

Product Code. 10772833 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10772833 5 g Glass bottle 5g
10492865 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10772833 Supplier Thermo Scientific Acros Supplier No. 369460050
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

Sodium glycocholate hydrate, CAS # 207614-05-9, is a bile acid glycine conjugate and also an ionic biologic detergent. | CAS: 207614-05-9 | C26H42NNaO6 | 487.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Sodium glycocholate hydrate is a conjugated bile salt
  • It is an ionic detergent

Applications

  • Sodium glycocholate hydrate is used to determine serum total bile acids by fluorimetric and enzymatic methods.
  • It has also been used in in vitro studies to investigate its effect on micelle formation and solubilization of testosterone.
  • This compound is considered a detergent used to solubilize fats for absorption.
  • This chemical product is synthetically derived from a natural compound, cholic acid.
  • Sodium glycocholate hydrate has been tested in the laboratory for its ability to form a liposomal delivery system to enhance oral insulin delivery. The in vitro results demonstrated that sodium glycocholate liposomes showed promising activity and can be useful to create a new approach to orally deliver insulin more efficiently.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 207614-05-9
Molecular Formula C26H42NNaO6
Molecular Weight (g/mol) 487.61
MDL Number MFCD09037360
InChI Key OABYVIYXWMZFFJ-XIMBWSGONA-M
Synonym glycocholic acid sodium salt hydrate, sodium glycocholate hydrate, c26h42no6.na.2h2o, sodium glycocholate dihydrate
PubChem CID 45051920
IUPAC Name sodium;2-[[(4R)-4-[(3R,5S,7R,8R,9S,10S,12S,13R,14S,17R)-3,7,12-trihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoyl]amino]acetate;dihydrate
SMILES [Na+].[H][C@@]1(CC[C@@]2([H])[C@]3([H])[C@H](O)C[C@]4([H])C[C@H](O)CC[C@]4(C)[C@@]3([H])C[C@H](O)[C@]12C)[C@H](C)CCC(=O)NCC([O-])=O

Specifications

Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 0.98
Packaging Glass bottle
Quantity 5 g
Specific Rotation Condition + 29.50 (20.00°C c=1,H2O)
Specific Rotation +28° to +31° (20°C, 589nm) (c=1, H2O) on dry basis
Formula Weight 487.61
Percent Purity ≥97.5% (on dry basis) (Total base)
Physical Form Liquid
Chemical Name or Material Sodium glycocholate hydrate, 0.98
Show More Show Less
compliance-icons
Product Identifier
  • Sodium glycocholate hydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.