missing translation for 'onlineSavingsMsg'
Learn More

Sodium glycolate, 97%

Product Code. 11439463
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11439463 100 g 100g
11449463 500 g 500g
11459463 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11439463 Supplier Thermo Scientific Alfa Aesar Supplier No. A12341.22
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 2836-32-0 | C2H3NaO3 | 98.033 g/mol

Sodium glycolate is used as a disintegrant, suspending agent, gelling agent, buffering agent and in pharmaceutical grade for tablets and capsules. Further, it is used in cosmetics and personal care products primarily as an exfoliant, as pH adjusters, skin-conditioning agents and as a flavoring agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium glycolate is used as a disintegrant, suspending agent, gelling agent, buffering agent and in pharmaceutical grade for tablets and capsules. Further, it is used in cosmetics and personal care products primarily as an exfoliant, as pH adjusters, skin-conditioning agents and as a flavoring agent.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2836-32-0
Molecular Formula C2H3NaO3
Molecular Weight (g/mol) 98.033
MDL Number MFCD00065358
InChI Key VILMUCRZVVVJCA-UHFFFAOYSA-M
Synonym sodium glycolate, sodium hydroxyacetate, sodium 2-hydroxyacetate, sodium lycolate, monosodium glycolate, glycolic acid sodium salt, natriumglykolat, sodium glycollate, acetic acid, hydroxy-, monosodium salt, glycolic acid, monosodium salt
PubChem CID 517347
IUPAC Name sodium;2-hydroxyacetate
SMILES C(C(=O)[O-])O.[Na+]

Specifications

Melting Point 210°C to 218°C
Assay Percent Range 97%
Linear Formula HOCH2CO2Na
Quantity 100 g
Solubility Information Soluble in water.
Formula Weight 98.03
Percent Purity 97%
Physical Form Powder or Lumps
Chemical Name or Material Sodium Glycolate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.