missing translation for 'onlineSavingsMsg'
Learn More

Sodium glycolate, 97%

Product Code. 10489661 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 10489661

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2836-32-0 | C2H3NaO3 | 98.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2836-32-0
Molecular Formula C2H3NaO3
Molecular Weight (g/mol) 98.03
MDL Number MFCD00065358
InChI Key VILMUCRZVVVJCA-UHFFFAOYSA-M
Synonym sodium glycolate, sodium hydroxyacetate, sodium 2-hydroxyacetate, sodium lycolate, monosodium glycolate, glycolic acid sodium salt, natriumglykolat, sodium glycollate, acetic acid, hydroxy-, monosodium salt, glycolic acid, monosodium salt
PubChem CID 517347
IUPAC Name sodium;2-hydroxyacetate
SMILES C(C(=O)[O-])O.[Na+]

Specifications

Color White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula NaOOCCH2OH
Quantity 100 g
Solubility Information Solubility in water: soluble.
Formula Weight 98.03
Percent Purity 97%
Physical Form Powder
Chemical Name or Material Sodium glycolate, 97%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.