missing translation for 'onlineSavingsMsg'
Learn More

Sodium hexabromoiridate(IV), Ir 24.1% min

Product Code. 11307497
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11307497

Brand: Thermo Scientific Alfa Aesar 012649.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 28529-99-9 | Br6IrNa2 | 717.621 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28529-99-9
Molecular Formula Br6IrNa2
Molecular Weight (g/mol) 717.621
MDL Number MFCD00064675
InChI Key XUAIOVIZUXDJGC-UHFFFAOYSA-H
Synonym sodium hexabromoiridate iv, disodium hexabromoiridiumdiuide, sodium hexabromoiridate iv , ir, iridate 2-,hexabromo-,sodium 1:2 , oc-6-11
PubChem CID 71311344
IUPAC Name disodium;hexabromoiridium(2-)
SMILES [Na+].[Na+].Br[Ir-2](Br)(Br)(Br)(Br)Br

Specifications

Melting Point >350°C
Physical Form Crystalline
Quantity 1 g
Formula Weight 717.63
Concentration or Composition (by Analyte or Components) Ir 24.1% min.
Sensitivity Hygroscopic
Chemical Name or Material Sodium hexabromoiridate(IV)

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.