missing translation for 'onlineSavingsMsg'
Learn More

Sodium hexabromoplatinate(IV) hexahydrate

Product Code. 11349359
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11349359

Brand: Thermo Scientific Alfa Aesar 088966.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 39277-13-9 | Br6Na2Pt | 720.488 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39277-13-9
Molecular Formula Br6Na2Pt
Molecular Weight (g/mol) 720.488
MDL Number MFCD00014237
InChI Key RFZKZIKSQSDBBI-UHFFFAOYSA-H
Synonym disodium hexabromoplatinate, sodium hexabromoplatinate iv, disodium hexabromoplatinumdiuide, platinate 2-, hexabromo-, disodium, oc-6-11
PubChem CID 20474473
IUPAC Name disodium;hexabromoplatinum(2-)
SMILES [Na+].[Na+].Br[Pt-2](Br)(Br)(Br)(Br)Br

Specifications

Physical Form Crystalline
Quantity 5 g
Formula Weight 828.49 (720.49 Anhydrous)
Chemical Name or Material Sodium hexabromoplatinate (IV) hexahydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.