missing translation for 'onlineSavingsMsg'
Learn More

Sodium hexacyanoferrate(II) decahydrate, TRC

Product Code. 30411319
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
Product Code. Quantity unitSize
30411319 5 g 5g
1 options
This item is not returnable. View return policy

Product Code. 30411319

Brand: Toronto Research Chemicals TRCS6343055G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sodium hexacyanoferrate(II) decahydrate
CAS 14434-22-1
Molecular Formula Na4Fe(CN)6 . 10(H2O)
Quantity 5 g
Synonym Sodium ferrocyanide decahydrate, Sodium ferrocyanide, Sodium hexacyanoferrate (Na4(Fe(CN)6))','decahydrate, Tetrasodium hexacyanoferrate(4-) decahydrate, Yellow prussiate of soda
InChI Formula InChI=1S/6CN.Fe.4Na.10H2O/c6*1-2;;;;;;;;;;;;;;;/h;;;;;;;;;;;10*1H2/q6*-1;+2;4*+1;;;;;;;;;;
SMILES O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[Na+].[Na+].[Fe+2].[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N
IUPAC Name tetrasodium;iron(2+);hexacyanide;decahydrate
Molecular Weight (g/mol) 303.91
Formula Weight 484.0181
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.