missing translation for 'onlineSavingsMsg'
Learn More

Sodium hexafluoroantimonate(V), 99.5%

Product Code. 11362158
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11362158

Brand: Thermo Scientific Alfa Aesar 013066.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 16925-25-0 | F6NaSb | 258.74 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16925-25-0
Molecular Formula F6NaSb
Molecular Weight (g/mol) 258.74
MDL Number MFCD00003482
InChI Key HKLMYZVMEYYVBS-UHFFFAOYSA-H
Synonym sodium hexafluoroantimonate, sodium hexafluoroantimonate v, sodium hexafluorostibanuide, sodium hexafluoro antimonate, antimony sodium fluoride, f6sb.na, nasbf6, acmc-20ajo7, sodium hexafluorostiboranuide, hexafluorostibine, sodium salt
PubChem CID 16689647
IUPAC Name sodium;hexafluoroantimony(1-)
SMILES F[Sb-](F)(F)(F)(F)F.[Na+]

Specifications

Color White
Physical Form Crystalline
Quantity 25 g
Assay Percent Range 99.5%
Formula Weight 258.73
Odor Odorless
Appearance White powder
Density 3.38
Chemical Name or Material Sodium hexafluoroantimonate(V)

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.