missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium hexafluorophosphate, 98.5+%, pure
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Plastic bottle
Unit Size:
10g
250g
50g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
Specifications
Specifications
| Melting Point | 200.0°C |
| Color | White to Gray |
| Physical Form | Crystalline Powder |
| Quantity | 250 g |
| Assay Percent Range | 98.5% min |
| Linear Formula | NaPF6 |
| Solubility Information | Solubility in water: soluble |
| Formula Weight | 167.95 |
| Percent Purity | ≥98.5% |
| Grade | Pure |
| Show More |
Safety and Handling
Product Identifier
- Sodium hexafluorophosphate
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction