missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hexametaphosphate, Pure, Flake, Mainly (NaPO3)6, Fisher Chemical™

Product Code. 10336340 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 g
1 kg
Unit Size:
1kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10336340 1 kg 1kg
10080480 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Product Code. 10336340 Supplier Fisher Chemical Supplier No. S/4120/60
  • ONLINE EXCLUSIVE PRICE
    27.10 €
    24.96 € / 1kg
    Save 2.14 €
    7% Off
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below

Na6O18P6, CAS Number-10124-56-8, 1kg, White, 611.7708g/mol, 640 deg.C, 136045, Plastic powder jar, 6.0 to 7.7, Solid

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10124-56-8
Molecular Formula Na6O18P6
Molecular Weight (g/mol) 611.77
MDL Number 136045

Specifications

Melting Point 640°C
Color White
pH 6.0 to 7.7
Physical Form Solid
Quantity 1 kg
Formula Weight 611.7708g/mol
Grade Pure
Packaging Plastic powder jar
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.