missing translation for 'onlineSavingsMsg'
Learn More

Sodium hexametaphosphate, tech.

Product Code. 11317018
Click to view available options
Quantity:
1 kg
5 kg
Unit Size:
1kg
5kg
This item is not returnable. View return policy

Product Code. 11317018

Brand: Thermo Scientific Alfa Aesar 014706.A7

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 10124-56-8 | Na6O18P6

Sodium hexametaphosphate is a food additive used in dairy product, canned milk, packaged egg whites, ice cream, seafood and meat processing. It is also used as a sequestrant and water softening agent and detergents. It is an active ingredient in toothpastes as an anti-staining agent. It prevents the steel corrosion. Further, it is used in building, water treatment and in photographic developing. In addition, it is used as a retarder in dental alginate impression materials and as a thinner for petroleum drilling fluids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 10124-56-8
Molecular Formula Na6O18P6
MDL Number MFCD00136045
Synonym Sodium metaphosphate

Especificaciones

Boiling Point 1500°C
Melting Point 628°C
Color White
Physical Form Granules
Quantity 5 kg
Solubility Information Soluble in water. Insoluble in organic solvents
Formula Weight 611.77
Grade Technical
Odor Odorless
Refractive Index 1.482
Appearance White granules
Chemical Name or Material Sodium hexametaphosphate
Mostrar más Mostrar menos

RUO – Research Use Only

Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado