missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hexanitrocobaltate(III), ACS Reagent, Honeywell™ Fluka™

Product Code. 15642320 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Poly Bottle
Unit Size:
100g
500g
Product Code. Quantity Packaging unitSize
15642320 100 g Poly Bottle 100g
15652320 500 g Poly Bottle 500g
2 options
This item is not returnable. View return policy

Product Code. 15642320

Brand: Honeywell-Fluka 233722100G

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ACS Reagent

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13600-98-1
Molecular Formula CoN6Na3O12
Molecular Weight (g/mol) 403.93
MDL Number MFCD00003512
InChI Key BZFWEBJDBATXME-UHFFFAOYSA-N
IUPAC Name trisodium hexanitrocobalttris(ylium)
SMILES [Na+].[Na+].[Na+].[O-][N+](=O)[Co+3]([N+]([O-])=O)([N+]([O-])=O)([N+]([O-])=O)([N+]([O-])=O)[N+]([O-])=O

Specifications

Melting Point 220°C
Color Golden or Orange-Yellow
Quantity 100 g
Linear Formula Na3Co(NO2)6
UN Number UN1479
Formula Weight 403.94g/mol
Grade ACS Reagent
Packaging Poly Bottle
Appearance Powder
Chemical Name or Material Sodium Hexanitrocobaltate(III)
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Sodium hexanitrocobaltate(III)
Signal Word
  • Warning
Hazard Category
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Oxidising solid Category 3
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Trisodium hexanitritocobaltate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.