missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hyaluronate, TRC

Product Code. 18092320
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18092320 25 g 25g
18011840 5 g 5g
18023630 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18092320 Supplier Toronto Research Chemicals Supplier No. TRCS63480025G
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9067-32-7
Molecular Formula [C14H22NNaO11]n
Molecular Weight (g/mol) 419.31
Synonym Arthrease, Artz, Artz Dispo, Artzal, Bio Hyaluro 12, Bio Sodium Hyaluronate HA 20, Chlamyhyaluronic acid sodium salt, Cystistat, Euflexxa, FCH 121-S, FCH 150, FCH 200, FCH 248, FCH 60, FCH 80, FCH 80LE, FCH-SU, Fermatron, HA-F, HA-Q, HA-Q 1, HA-QA, HE-QSE, Healon, Healon (polysaccharide), Healon GV, Healon V, Hyalart, Hyalein, Hyalein Mini, Hyalgan, Hyaluronsan HA-LQ, Hyaluronsan HA-LQ1, Hyaluronsan HA-LQH, Hyaluronsan HA-Q, Hyaluronsan HA-QSS, Hyaluronsan M 5070, Hyasol, Hyasol BT, Hyladerm, Hylo Comod, Jointex, Khionat, NRD 101, Nidelon, Opegan, Orthovisc, Ostenil, Provisc, SI 4402, SL 1010, SLM 10, SPH, Sinovial, Sodium hyaluronate, Suvenyl, Tear Balance, Vismed
IUPAC Name sodium (2S,3S,5R,6S)-3-(((2S,3R,4R,5S,6R)-3-acetamido-5-hydroxy-6-(hydroxymethyl)-4-methoxytetrahydro-2H-pyran-2-yl)oxy)-4,5-dihydroxy-6-methyltetrahydro-2H-pyran-2-carboxylate
SMILES C[C@@H]([C@@H](C([C@@H]1O[C@H]2[C@@H]([C@H]([C@@H]([C@H](O2)CO)O)OC)NC(C)=O)O)O)O[C@@H]1C(O[Na])=O

Specifications

Quantity 25 g
InChI Formula InChI=1S/C16H27NO11.Na/c1-5-9(20)11(22)13(14(26-5)15(23)24)28-16-8(17-6(2)19)12(25-3)10(21)7(4-18)27-16;/h5,7-14,16,18,20-22H,4H2,1-3H3,(H,17,19)(H,23,24);/q;+1/p-1/t5-,7+,8+,9-,10+,11?,12+,13-,14-,16-;/m0./s1
Formula Weight 419.104
Chemical Name or Material Hyaluronic Acid Sodium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.