missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium hydride, 60% dispersion in mineral oil, in soluble bags
Shop All Thermo Scientific Chemicals ProductsDescription
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Adsorbent, Reduction reagent
Specifications
Specifications
| Melting Point | 800.0°C |
| Color | Gray to Yellow |
| Physical Form | Tacky Powder |
| Quantity | 100 g |
| Linear Formula | NaH |
| Fieser | 02,382; 01,1075; 04,452; 05,610; 06,541; 07,335; 08,458; 09,427; 11,486; 12,447; 14,288; 16,307 |
| Flash Point | 165°C |
| Merck Index | 15, 8760 |
| Solubility Information | Solubility in water: reacts. Other solubilities: reacts with alcohols, soluble in molten sodium hydroxide, insoluble in cs2, ccl4, liquid nh3, benzene |
| Formula Weight | 24 |
| Show More |
Safety and Handling
Product Identifier
- Sodium hydride, 60% dispersion in mineral oil
- Danger
- Substance which\, in contact with water\, emits flammable gases Category 1
- H260-In contact with water releases flammable gases which may ignite spontaneously.
- MIXTURE LIST-Contains : Sodium hydride
RUO – Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?