missing translation for 'onlineSavingsMsg'
Learn More

Sodium hydride, 60% dispersion in mineral oil, in soluble bags

Product Code. 10646772 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
250 g
1 kg
Unit Size:
100g
1kg
250g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10646772 250 g 250g
10104210 100 g 100g
10367270 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 10646772 Supplier Thermo Scientific Acros Supplier No. 189862500
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 7646-69-7 | HNa | 24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Adsorbent, Reduction reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7646-69-7 , 8042-47-5
Molecular Formula HNa
Molecular Weight (g/mol) 24
MDL Number MFCD00003471
InChI Key BZKBCQXYZZXSCO-UHFFFAOYSA-N
Synonym sodium hydride, sodiumhydride, sodium hydride nah, sodium hydride oil dispersion, sodium monohydride, sodium hydride nad, sodiumhydrid, natrium hydride, sodium hydrid
PubChem CID 24758
IUPAC Name sodium;hydride
SMILES [H-].[Na+]

Specifications

Melting Point 800.0°C
Color Gray to Yellow
Physical Form Tacky Powder
Quantity 250 g
Linear Formula NaH
Fieser 02,382; 01,1075; 04,452; 05,610; 06,541; 07,335; 08,458; 09,427; 11,486; 12,447; 14,288; 16,307
Flash Point 165°C
Merck Index 15, 8760
Solubility Information Solubility in water: reacts. Other solubilities: reacts with alcohols, soluble in molten sodium hydroxide, insoluble in cs2, ccl4, liquid nh3, benzene
Formula Weight 24
Percent Purity 57 to 63% (NaH)
Density 1.2000g/mL
Chemical Name or Material Sodium hydride, 60% dispersion in mineral oil
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.