missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydrogen Carbonate, Extra Pure, SLR, Fisher Chemical™

Product Code. 10508640 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 kg
3 kg
5 kg
Packaging:
Plastic keg
Plastic powder jar
Unit Size:
1kg
3kg
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10508640 1 kg Plastic powder jar 1kg
10010510 3 kg Plastic powder jar 3kg
10122640 5 kg Plastic keg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10508640 Supplier Fisher Chemical Supplier No. S/4200/60
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Sodium Hydrogen Carbonate, Extra Pure, SLR

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 144-55-8
Molecular Formula CHNaO3
Molecular Weight (g/mol) 84.01
MDL Number MFCD00003528
InChI Key UIIMBOGNXHQVGW-UHFFFAOYSA-M
Synonym sodium bicarbonate, sodium hydrogen carbonate, baking soda, carbonic acid monosodium salt, sodium acid carbonate, bicarbonate of soda, sodium hydrogencarbonate, meylon, acidosan, neut
PubChem CID 516892
ChEBI CHEBI:32139
SMILES [Na+].OC([O-])=O

Specifications

Melting Point 270°C
Color White
pH 8.3
Physical Form Powder/Solid
Quantity 1 kg
IUPAC Name sodium;hydrogen carbonate
Formula Weight 84.01g/mol
Packaging Plastic powder jar
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.