missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydrogen Sulfate, TRC
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | Sodium Hydrogen Sulfate |
| CAS | 7681-38-1 |
| Molecular Formula | NaHSO4 |
| Quantity | 5 g |
| Synonym | Acid Sodium Sulfate, BIF, Bisulfate of Soda, Fanal, Monobasic Sodium Sulfate, Monosodium Hydrogen Sulfate, Monosodium Sulfate, Niter cake, Nitre cake, Poultry Litter Treatment, Printgen N, SBS Pet, Sodium Acid Sulfate, Sodium Bisulfate, Sodium Bisulphate, Sodium Hydrogen Sulfate, Sodium Hydrogen Sulfate (NaHSO4), Sodium Hydrosulfate, WC 00, WC-Klosettreiniger, WC-Perfect, WC-Super, pHase (Sodium Bisulfite) |
| InChI Formula | InChI=1S/Na.H2O4S/c;1-5(2,3)4/h;(H2,1,2,3,4)/q+1;/p-1 |
| SMILES | [Na+].OS(=O)(=O)[O-] |
| IUPAC Name | sodium;hydrogen sulfate |
| Molecular Weight (g/mol) | 120.06 |
| Formula Weight | 119.9493 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction