missing translation for 'onlineSavingsMsg'
Learn More

Sodium hydrogen tartrate, 98+%, for analysis

Product Code. 10453005 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10453005 250 g Plastic bottle 250g
10443005 1 kg Plastic bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10453005 Supplier Thermo Scientific Acros Supplier No. 389422500
  • ONLINE EXCLUSIVE PRICE
    57.40 €
    55.70 € / 250g
    Save 1.70 €
    2% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 526-94-3 | C4H5NaO6 | 172.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 526-94-3
Molecular Formula C4H5NaO6
Molecular Weight (g/mol) 172.07
MDL Number MFCD00065393,MFCD00065393
InChI Key NKAAEMMYHLFEFN-UHFFFAOYNA-M
Synonym sodium bitartrate, monosodium tartrate, sodium hydrogen tartrate, natriumtartrat german, sodium 3-carboxy-2,3-dihydroxypropanoate, monobasic sodium tartrate, monosodium l-+-tartrate, tartaric acid, monosodium salt, natrium hydrogen-2r,3r-tartrat, weinstein
PubChem CID 23690454
IUPAC Name sodium;2,3,4-trihydroxy-4-oxobutanoate
SMILES [Na+].OC(C(O)C([O-])=O)C(O)=O

Specifications

Melting Point 253°C
Color White
pH 3.3 to 3.6 (5% soln.)
Physical Form Powder
Quantity 250 g
Assay Percent Range 98+%
Merck Index 15, 8725
Solubility Information Solubility in water: 89g/L (20°C)
Formula Weight 172.07
Percent Purity ≥98%
Grade Analytical
Packaging Plastic bottle
Chemical Name or Material Sodium hydrogen tartrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.