missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydrogen Tartrate Monohydrate, Certified AR for Analysis, Fisher Chemical

Product Code. 11473723 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Plastic powder jar
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11473723 500 g Plastic powder jar 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11473723 Supplier Fisher Chemical Supplier No. S/4760/53
Quantity Price
1 209.48 € / 500g
4 188.54 € / 500g

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

500g, Plastic powder jar

TRUSTED_SUSTAINABILITY

Specifications

CAS 6131-98-2
Color White
Physical Form Solid
Quantity 500 g
Molecular Formula C4H7NaO7
MDL Number 211332
Synonym sodium bitartrate monohydrate, sodium hydrogen tartrate monohydrate, sodium bitartrate,monohydrate, tartaric acid monosodium salt, sodium 3-carboxy-2,3-dihydroxypropanoate hydrate, l-+-tartaric acid monosodium salt, sodium bitartrate, monohydrate,, sodium bitartrate monohydrate, bioultra t, sodium 3-carboxy-2,3-dihydroxypropanoate-water 1/1/1, sodium bitartrate monohydrate, purum p.a t
InChI Key LLVQEXSQFBTIRD-UHFFFAOYSA-M
SMILES C(C(C(=O)[O-])O)(C(=O)O)O.O.[Na+]
IUPAC Name sodium;2,3,4-trihydroxy-4-oxobutanoate;hydrate
Molecular Weight (g/mol) 190.083
PubChem CID 23678966
Formula Weight 190.09g/mol
Packaging Plastic powder jar
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.