missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydrogencitrate Sesquihydrate, TRC

Product Code. 30420643
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30420643 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30420643 Supplier Toronto Research Chemicals Supplier No. TRCS61521550G
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sodium Hydrogencitrate Sesquihydrate
CAS 6132-05-4
Molecular Formula C12H12Na4O14 3(H2O)
Quantity 50 g
Synonym 1,2,3-Propanetricarboxylic acid','2-hydroxy-','sodium salt','hydrate (2:4:3), 1,2,3-Propanetricarboxylic acid','2-hydroxy-','disodium salt','hydrate (2:3) (9CI), Citric acid','disodium salt','sesquihydrate (8CI), Sodium 1,2,3-propanetricarboxylate hydrate (4:2:3), Disodium hydrogen citrate 1.5 hydrate, Disodium citrate sesquihydrate, Tetrasodium dicitrate trihydrate
InChI Formula InChI=1S/2C6H8O7.4Na.3H2O/c2*7-3(8)1-6(13,5(11)12)2-4(9)10;;;;;;;/h2*13H,1-2H2,(H,7,8)(H,9,10)(H,11,12);;;;;3*1H2/q;;4*+1;;;/p-4
SMILES O.O.O.[Na+].[Na+].[Na+].[Na+].OC(=O)C(O)(CC(=O)[O-])CC(=O)[O-].OC(=O)C(O)(CC(=O)[O-])CC(=O)[O-]
IUPAC Name tetrasodium;3-carboxy-3-hydroxypentanedioate;trihydrate
Molecular Weight (g/mol) 526.22
Formula Weight 526.0135
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.