missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydrosulfide Hydrate, Honeywell Fluka™

Product Code. 15691530 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
5 g
100 g
1 kg
3 kg
Packaging:
Glass Bottle
Unit Size:
100g
1kg
3kg
5g
This item is not returnable. View return policy

Product Code. 15691530

Brand: Honeywell-Fluka 1615271KG

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

min. 60.0% (NaHS)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 207683-19-0
Molecular Formula HNaS
Molecular Weight (g/mol) 56.06
MDL Number MFCD00149796
InChI Key HYHCSLBZRBJJCH-UHFFFAOYSA-M
Synonym sodium hydrosulfide hydrate, hnas.h2o, nahs.h2o, sodium sulfanide hydrate, sodium bisulfide monohydrate, sodium hydrosulphide hydrate, sodium hydro sulfide hydrate, sodium hydrosulfide monohydrate, sodium hydrate sulfanide, sodium hydrosulfide hydrate, cp
PubChem CID 16211346
SMILES [Na+].[SH-]

Specifications

Melting Point 52°C to 54°C
Color Yellow
Quantity 1 kg
Linear Formula NaSH·xH2O
UN Number UN2949
Flash Point 90°C (194°F)
IUPAC Name sodium sulfanide
Formula Weight 56.06g/mol (anhydrous basis)
Packaging Glass Bottle
Appearance Flakes
Chemical Name or Material Sodium Hydrosulfide Hydrate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.