missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydrosulfite (85%), TRC

Product Code. 30427869
Change view
Click to view available options
Quantity:
250 g
Unit Size:
250g
Product Code. Quantity unitSize
30427869 250 g 250g
1 options
This item is not returnable. View return policy

Product Code. 30427869

Brand: Toronto Research Chemicals TRCS634810250G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sodium Dithionite
CAS 7775-14-6
Molecular Formula 2 Na . O4 S2
Quantity 250 g
Synonym Dithionous acid','sodium salt (1:2), Dithionous acid','disodium salt (8CI,9CI), Blankit, Blankit IN, Blankit S, Burmol, Cyclanon ARC, Disodium dithionite, Disodium hydrosulfite, Hydros, Hydrosulfite R Conc, Sodium dithionite, Sodium dithionite (Na2(S2O4)), Sodium hydrosulfite, Sodium hydrosulfite (Na2S2O4), Sodium hyposulfite, V-Brite B, Vatrolite
InChI Formula InChI=1S/2Na.H2O4S2/c;;1-5(2)6(3)4/h;;(H,1,2)(H,3,4)/q2*+1;/p-2
SMILES [Na+].[Na+].[O-]S(=O)S(=O)[O-]
Molecular Weight (g/mol) 174.11
Formula Weight 173.9033
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.