missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium hydrosulfite, ca. 85%, Tech.
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
1 kg
2 kg
Packaging:
Glass bottle
Unit Size:
1kg
25g
2kg
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
Specifications
Specifications
| Melting Point | >300.0°C |
| Color | White |
| Physical Form | Crystalline Powder |
| Quantity | 25 g |
| Assay Percent Range | ca. 85% |
| Molecular Formula | Na2O4S2 |
| Linear Formula | Na2S2O4 |
| Merck Index | 15, 8747 |
| Solubility Information | Solubility in water: 250g/L (20°C). Other solubilities: slightly soluble in alcohol |
| Molecular Weight (g/mol) | 174.1 |
| Show More |
Safety and Handling
Product Identifier
- Sodium hydrosulfite
- Danger
- Acute toxicity Category 4
- Self-heating substance Category 1
- H302-Harmful if swallowed.
- H251-Self-heating: may catch fire.
- EUH031-Contact with acids liberates toxic gas.
- P235+P410-Keep cool. Protect from sunlight.
- P264-Wash thoroughly after handling.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction