missing translation for 'onlineSavingsMsg'
Learn More

Sodium hydrosulfite, tech., 85+%

Product Code. 11317708
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
Unit Size:
100g
1kg
25g
Product Code. Quantity unitSize
11317708 25 g 25g
11307708 100 g 100g
11397698 1 kg 1kg
3 options
This item is not returnable. View return policy

Product Code. 11317708

Brand: Thermo Scientific Alfa Aesar 033381.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7775-14-6 | Na2O4S2 | 174.096 g/mol

Sodium hydrosulfite is used as a reducing agent in aqueous solutions, sulfonating agent , chelating agent and decolorizing agent in organic reactions. It finds application in water treatment, gas purification, cleaning, leather, polymers, photography, and many others. It is involved in chemical enhanced oil recovery to stabilize polyacrylamide polymers against radical degradation in the presence of iron. It plays an important role to determine the iron content in soil chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7775-14-6
Molecular Formula Na2O4S2
Molecular Weight (g/mol) 174.096
MDL Number MFCD00011640
InChI Key JVBXVOWTABLYPX-UHFFFAOYSA-L
Synonym sodium dithionite, sodium hydrosulfite, disodium dithionite, vatrolite, sodium sulfoxylate, sodium hydrosulphite, dithionous acid, disodium salt, blankit, burmol, hydros
PubChem CID 24489
ChEBI CHEBI:66870
SMILES [O-]S(=O)S(=O)[O-].[Na+].[Na+]

Specifications

Melting Point 52°C (decomposition)
Color White
Physical Form Powder
Quantity 25 g
Assay Percent Range >85%
Solubility Information Soluble in water. Slightly soluble in alcohol.
Formula Weight 174.11
Sensitivity Moisture sensitive
Appearance White powder
Chemical Name or Material Sodium hydrosulfite
compliance-icons compliance-icons
Product Identifier
  • Sodium hydrosulfite, technical grade
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Self-heating substance Category 1
Hazard Statement
  • H251-Self-heating: may catch fire.
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
  • EUH031-Contact with acids liberates toxic gas.
Precautionary Statement
  • P235+P410-Keep cool. Protect from sunlight.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Sodium dithionite

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.