missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, 0.02M, Analytic Volumetric Solution, Reagecon™
BIOPHARMA

Product Code. 11592982 Shop All Reagecon Products
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11592982 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11592982 Supplier Reagecon Supplier No. S20021
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Provides a convenient method for accurate volumetric analysis in the laboratory. Reagecon Sodium Hydroxide 1.0M, Low in Carbonate, Analytical Volumetric Solution is a cost-effective answer to time- and labor-consuming preparation problems.

Ready to use aqueous volumetric solution for all analytical laboratory applications, which can be used for manual or automatic titrations. This product is certified to a specification as outlined in the Certificate of Analysis. Lot Number and Expiry Date are printed on the label. Solution is certified traceable to N.I.S.T and this will be stated in the Certificate of Analysis.

  • Molecular Formula: NaOH
  • Molar Mass: 40.00g/mol
  • CAS: 1310-73-2
  • EIN ECS No.: 215-185-5

Specifications

Color Colorless
Boiling Point 100°C
CAS 1310-73-2
Melting Point 0°C
Molecular Formula HNaO
Concentration 0.02mol/L
Density 1.00091g/cm3
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
Solubility Information Fully miscible
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Quantity 1 L
SMILES [OH-].[Na+]
IUPAC Name sodium;hydroxide
Molecular Weight (g/mol) 39.997
PubChem CID 14798
ChEBI CHEBI:32145
Formula Weight 40.00g/mol
Normality 0.02N
Traceability to NIST Traceable to NIST
Vapor Pressure 23hPa (at 20°C)
Physical Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.