missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide 0.313 M For Laboratory Use and Titrations, Chem Lab

Product Code. 15943947
Change view
Click to view available options
Sodium Hydroxide 0.313 M For Laboratory Use and Titrations, 2.5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
15943947 Sodium Hydroxide 0.313 M For Laboratory Use and Titrations, 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15943947 Supplier Chem-Lab Supplier No. CL05.1456.2500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Color Colorless
Boiling Point 100°C
CAS 1310-73-2
Melting Point 0°C
Concentration 0.313 M
Density 1.02 g/mL
Quantity 2.5 L
Molecular Weight (g/mol) 40
Packaging Polyethylene
Traceability to NIST Reference standards acc. NIST SRM 723e
Physical Form Liquid
Show More Show Less
compliance-icons
Product Identifier
  • Sodium hydroxide 0.313 mol/l
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H290-May be corrosive to metals.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: sodium hydroxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.