missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, 1M, BAKER ANALYZED, J.T. Baker™

Product Code. 10026070 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
10 L
Packaging:
Polycube (HDPE)
Unit Size:
10 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10026070 10 L Polycube (HDPE) 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10026070 Supplier JT Baker Supplier No. 7097.9010

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High quality grade volumetric solution is ready to use for laboratory and specialized industrial applications.

  • Standardized potentiometrically against an NIST Standard Reference Material, Potassium Hydrogen Phthalate
  • Label provides an actual Lot Analysis as a guarantee of purity and consistency
  • Analytical methods used for these determinations are based on standard test procedures described in technical publications
  • Improvements developed by J.T. Baker are employed to achieve a higher degree of reliability

Specifications

Color Colorless
Boiling Point 100°C
CAS 1310-73-2
Melting Point 0°C
Molecular Formula HNaO
Concentration 0.997–1.003mol/L
Density 1g/cm3
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
Solubility Information Fully miscible in water
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Quantity 10 L
SMILES [OH-].[Na+]
IUPAC Name sodium;hydroxide
Molecular Weight (g/mol) 39.997
PubChem CID 14798
ChEBI CHEBI:32145
Formula Weight 39.997g/mol
Grade Standardized potentiometrically against an NIST Standard Reference Material, Potassium Hydrogen Phthalate
Normality 1N
Odor Odorless
Packaging Polycube (HDPE)
Physical Form Liquid
Chemical Name or Material Sodium Hydroxide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.