missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, 1M, Eu. Ph., Chem-Lab

Product Code. 13228323
Change view
Click to view available options
Quantity:
1 L
Packaging:
PE
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
13228323 1 L PE 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13228323 Supplier Chem-Lab Supplier No. CL07.1427.1000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For laboratory use. Meets specifications of Eur. Ph. 4(4.2.2) - (3006300).

Specifications

CAS 1310-73-2
Molecular Formula HNaO
Concentration 40g NaOH / 1L H2O
UN Number UN1824
Density 1.04g/mL
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Quantity 1 L
SMILES [OH-].[Na+]
IUPAC Name sodium;hydroxide
Molecular Weight (g/mol) 39.997
PubChem CID 14798
ChEBI CHEBI:32145
Formula Weight 40.00g/mol
Normality 1N
Packaging PE
Chemical Name or Material Sodium hydroxide
Show More Show Less
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Sodium hydroxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.