missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, 1N Volumetric Solution, BAKER ANALYZED™ Reagent, J.T. Baker™

Product Code. 15568344 Shop All J.T.Baker Products
Click to view available options
Quantity:
1 L
4 L
Packaging:
Cubetainer
Narrow Mouth Poly Bottle
Unit Size:
4 L
Each
This item is not returnable. View return policy

Product Code. 15568344

Brand: Avantor J.T.Baker 563503

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

High quality chemicals for laboratory and specialized industrial use

Chemical Identifiers

CAS 1310-73-2
Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]

Specifications

Quantity 4 L
Formula Weight 40
Grade BAKER ANALYZED™ Reagent
Packaging Cubetainer
Specific Gravity 1L = 1.05kg
Chemical Name or Material Sodium Hydroxide, 1N Volumetric Solution
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.