missing translation for 'onlineSavingsMsg'
Learn More

Sodium hydroxide 40% in aqueous solution, BAKER, J.T. Baker™

Product Code. 10578342 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
10 L
Unit Size:
10 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10578342 10 L 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10578342 Supplier JT Baker Supplier No. 6165.9010
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Laboratory Reagent

Specifications

Boiling Point 119°C to 135°C (1013 hPa)
CAS 1310-73-2
Melting Point 9°C to 10°C
Assay Percent Range min. 40.0%
Molecular Formula HNaO
MDL Number MFCD00003548
Density min. 1.43 g/mL (at 20°C)
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Quantity 10 L
SMILES [OH-].[Na+]
IUPAC Name sodium hydroxide
Molecular Weight (g/mol) 40.00
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.