missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, 5N Volumetric Solution, BAKER ANALYZED™ Reagent, J.T. Baker™

Product Code. 15177153 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
1 L
4 L
Packaging:
Cubetainer
Narrow Mouth Poly Bottle
Unit Size:
1 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15177153 1 L Narrow Mouth Poly Bottle 1 L
15578354 4 L Cubetainer 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15177153 Supplier Avantor J.T.Baker Supplier No. 567102

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High quality chemicals for laboratory and specialized industrial use

Chemical Identifiers

CAS 1310-73-2
Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]

Specifications

Quantity 1 L
Formula Weight 40
Grade BAKER ANALYZED™ Reagent
Packaging Narrow Mouth Poly Bottle
Specific Gravity 1L = 1.17kg
Chemical Name or Material Sodium Hydroxide, 5N Volumetric Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.