missing translation for 'onlineSavingsMsg'
Learn More

Sodium hydroxide, 98.5%, for analysis, pellets

Product Code. 10743591 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 kg
2.5 kg
10 kg
25 kg
Packaging:
Fibre drum
Plastic Bottle
Plastic drum
Unit Size:
10kg
1kg
2.5kg
25kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10743591 1 kg Plastic Bottle 1kg
10338630 2.5 kg Plastic Bottle 2.5kg
10104473 10 kg Plastic drum 10kg
13408458 25 kg Fibre drum 25kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10743591 Supplier Thermo Scientific Acros Supplier No. 206060010
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1310-73-2 | HNaO | 39.997 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1310-73-2
Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
MDL Number MFCD00003548
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]

Specifications

Color White
Boiling Point 1390.0°C
Melting Point 318.0°C
Assay Percent Range 98.5%
Linear Formula NaOH
Fieser 01,1083; 05,616; 07,336; 08,460
Density 2.13g/mL
Solubility Information Solubility in water: completely soluble. Other solubilities: soluble in alcohol and glycerol, insoluble in acetone
Quantity 1 kg
Formula Weight 40
Percent Purity 98.5%
Grade Analytical
Packaging Plastic Bottle
Physical Form Pellet
Chemical Name or Material Sodium hydroxide
Show More Show Less
compliance-icons
Product Identifier
  • Sodium hydroxide
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.