missing translation for 'onlineSavingsMsg'
Learn More

Sodium hydroxide, 98.5%, for analysis, pellets

Product Code. 13408458 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 kg
2.5 kg
10 kg
25 kg
Packaging:
Fibre drum
Plastic Bottle
Plastic drum
Unit Size:
10kg
1kg
2.5kg
25kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
13408458 25 kg Fibre drum 25kg
10743591 1 kg Plastic Bottle 1kg
10338630 2.5 kg Plastic Bottle 2.5kg
10104473 10 kg Plastic drum 10kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 13408458 Fournisseur Thermo Scientific Acros Code fournisseur 206060250
  • ONLINE EXCLUSIVE PRICE
    398.00 €
    386.00 € / 25kg
    Save 12.00 €
    3% Off
 Restricted Item
Estimated Shipment: 27-10-2026
Ajouter au panier
Ajouter au panier
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

CAS: 1310-73-2 | HNaO | 39.997 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 1310-73-2
Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
MDL Number MFCD00003548
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]

Spécification

Color White
Boiling Point 1390.0°C
Melting Point 318.0°C
Assay Percent Range 98.5%
Linear Formula NaOH
Fieser 01,1083; 05,616; 07,336; 08,460
Solubility Information Solubility in water: completely soluble. Other solubilities: soluble in alcohol and glycerol, insoluble in acetone
Quantity 25 kg
Formula Weight 40
Percent Purity 98.5%
Grade Analytical
Packaging Fibre drum
Physical Form Pellet
Chemical Name or Material Sodium hydroxide
Afficher plus Afficher moins
compliance-icons
Identifiant produit
  • Sodium hydroxide
Mention d'avertissement
  • Danger
Catégorie de danger
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
Mention de danger
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Conseils de prudence
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Informations supplémentaires
  • MIXTURE LIST-Contain: Sodium hydroxide

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.