missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, 99%, BAKER ANALYZED, J.T. Baker™

Product Code. 10315920 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
5 kg
Packaging:
HDPE
Unit Size:
5kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10315920 5 kg HDPE 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10315920 Supplier JT Baker Supplier No. 0403.5000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High quality grade chemical for laboratory and specialized industrial use.

  • Label provides an actual Lot Analysis as a guarantee of purity and consistency
  • Analytical methods used for these determinations are based on standard test procedures described in technical publications
  • Improvements developed by J.T. Baker are employed to achieve a higher degree of reliability

Specifications

Color White
Boiling Point 1390°C
CAS 1310-73-2
Melting Point 319°C
Molecular Formula HNaO
Density 2.13g/cm3 at 20°C
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
Solubility Information 420g/L H2O
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Quantity 5 kg
SMILES [OH-].[Na+]
IUPAC Name sodium;hydroxide
Molecular Weight (g/mol) 39.997
PubChem CID 14798
ChEBI CHEBI:32145
Formula Weight 39.997g/mol
Percent Purity ≥99.0%
Grade BAKER ANALYZED
Odor Odorless
Packaging HDPE
Physical Form Solid
Chemical Name or Material Sodium Hydroxide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.