missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, Certified AR for Analysis, 30% Solution, d=1.33, Fisher Chemical™

Product Code. 10060622 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
5 L
Unit Size:
1 L
2.5 L
5 L
Product Code. Quantity unitSize
10060622 1 L 1 L
10263912 2.5 L 2.5 L
15960957 5 L 5 L
3 options
This item is not returnable. View return policy

Product Code. 10060622

Brand: Fisher Chemical S/4950/PB15

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 1310-73-2
Molecular Formula HNaO
MDL Number 3548
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
PubChem CID 14798
ChEBI CHEBI:32145
SMILES [OH-].[Na+]

Specifications

pH 12
Color Colorless
Boiling Point 120°C
Melting Point 8°C
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
Quantity 1 L
IUPAC Name sodium;hydroxide
Molecular Weight (g/mol) 39.997
Packaging HDPE Plastic Bottle
Physical Form Liquid
compliance-icons
Product Identifier
  • Sodium hydroxide, 30% solution, d=1,33
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Sodium hydroxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.