missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, Certified AR for Analysis, Pellets, meets analytical specification of Ph.Eur., BP, Fisher Chemical™

Product Code. 10538260 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 g
1 kg
3 kg
5 kg
Unit Size:
1kg
3kg
500g
5kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10538260 5 kg 5kg
10396240 500 g 500g
10675692 1 kg 1kg
10254540 3 kg 3kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 10538260 Leverandør Fisher Chemical Leverandørnr. S/4920/65
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

HNaO, CAS Number-1310-73-2, aetznatron, ascarite, caustic soda, soda, caustic, soda lye, sodium hydrate, sodium hydroxide, sodium hydroxide na oh, sodium hydroxide solution, white caustic, 1kg, 1390 deg.C, CHEBI:32145, White, 40g/mol, HEMHJVSKTPXQMS-UHFFFAOYSA-M, sodium;hydroxide, 318 deg.C, 3548 | CAS: 1310-73-2 | HNaO | 39.997 g/mol

HNaO, CAS Number-1310-73-2, white caustic, aetznatron, sodium hydroxide na oh, caustic soda, ascarite, soda lye, soda, caustic, sodium hydroxide solution, sodium hydrate, sodium hydroxide, 5kg, 1390 deg.C, CHEBI:32145, White, 40g/mol, HEMHJVSKTPXQMS-UHFFFAOYSA-M, sodium;hydroxide, 318 deg.C, 3548

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 1310-73-2
Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
MDL Number 3548
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]

Tekniske data

pH 14
Color White
Boiling Point 1390°C
Melting Point 318°C
Quantity 5 kg
Formula Weight 40g/mol
Packaging Plastic powder jar
Vapor Pressure 1mbar 700
Physical Form Solid
compliance-icons
Produktidentifikator
  • Sodium hydroxide
Signalord
  • Danger
Farekategori
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
Faresætning
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
Sikkerhedssætning
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplerende oplysninger
  • MIXTURE LIST-Contain: Sodium hydroxide
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.