missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide Certified AR, Fisher Chemical™

Product Code. 11918033
Change view
Click to view available options
Quantity: 1L, Packaging: Plastic
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
11918033 Quantity: 1L, Packaging: Plastic
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11918033 Supplier Fisher Bioreagents Supplier No. USSS2541
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sodium hydroxide 50% w/w solution 1L Certified AR suitable Dionex | CAS: 1310-73-2

Sodium hydroxide 50% w/w solution 1L Certified AR suitable Dionex

  • Sodium hydroxide 50% w/w solution 1L Certified AR suitable Dionex Plastic Bottle Cas No. 1310-73-2
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sodium hydroxide
Boiling Point 145°C
CAS 1310-73-2
Melting Point 681°C
Quantity 1 L
Percent Purity 50%
Packaging PLASTIC
compliance-icons
Product Identifier
  • Sodium hydroxide
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: sodium hydroxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.