missing translation for 'onlineSavingsMsg'
Learn More

Sodium hydroxide monohydrate, 99.996% (metals basis)

Product Code. 11359667
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11359667 25 g 25g
11369667 5 g 5g
11379667 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11359667 Supplier Thermo Scientific Alfa Aesar Supplier No. 041281.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12179-02-1 | H3NaO2 | 58.01 g/mol

Sodium hydroxide monohydrate is used as an acid neutralizer. It is a strong base widely used in the chemical industry to drive chemical reactions. It finds application as a neutralizing agent in the petroleum refining industry, as paint stripper and an analytical reagent in laboratories to measure the concentration of acids by titration. It is involved in soap production, alumina production by Bayer process, paper industry. It plays an important role as a catalyst for the transesterification of methanol and triglycerides in order to prepare biodiesel.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 12179-02-1
Molecular Formula H3NaO2
Molecular Weight (g/mol) 58.01
MDL Number MFCD00149229
InChI Key JBJWASZNUJCEKT-UHFFFAOYSA-M
Synonym sodium hydroxide monohydrate, na.ho.h2o, naoh water, water naoh, sodium hydroxide h2o, sodium hydroxide water, water sodium hydroxide, h2o naoh, naoh h2o, sodium,hydroxide,hydrate
PubChem CID 23719242
SMILES O.[OH-].[Na+]

Specifikationer

Color White
Boiling Point 1390°C
Melting Point 318.4°C
Assay Percent Range 99.996% (metals basis)
Density 2.13 g/cm3 at 20°C
Solubility Information Soluble in water and methanol.
Quantity 25 g
IUPAC Name sodium;hydroxide;hydrate
Formula Weight 58.01 (40.00 Anhydrous)
Odor Odorless
Physical Form Crystals
Chemical Name or Material Sodium hydroxide monohydrate
Visa mer Visa mindre
compliance-icons
Produktbeteckning
  • Sodium hydroxide monohydrate
Signalord
  • Danger
Riskkategorier
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A
Faroangivelser
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Skyddsangivelse
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Kompletterande information
  • MIXTURE LIST-Contain: Sodium hydroxide monohydrate

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.