missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide On Carrier, p.a., Honeywell™

Product Code. 15630680 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
250 g
Packaging:
Poly Bottle
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15630680 250 g Poly Bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15630680 Supplier Honeywell Research Chemicals Supplier No. 03350250G
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

p.a., for elemental analysis

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
    Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
    Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
TRUSTED_SUSTAINABILITY

Specifications

pH Alkaline
Color Black
Boiling Point 1390°C
CAS 1310-73-2
Melting Point 319°C
Molecular Formula HNaO
UN Number UN1823
Density 2.13g/cu.cm at 20°C
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Quantity 250 g
SMILES [OH-].[Na+]
IUPAC Name sodium hydroxide
Molecular Weight (g/mol) 40.00
Grade p.a.
Packaging Poly Bottle
Chemical Name or Material Sodium Hydroxide On Carrier
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.