missing translation for 'onlineSavingsMsg'
Learn More

Sodium hydroxide, 1N standard solution

Product Code. 10706341 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 L
5 L
25 L
Packaging:
Plastic Bottle
Plastic Can
Plastic drum
Unit Size:
1 L
25 L
5 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10706341 1 L Plastic Bottle 1 L
10784074 5 L Plastic drum 5 L
11974131 25 L Plastic Can 25 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10706341 Supplier Thermo Scientific Acros Supplier No. 124260010
  • ONLINE EXCLUSIVE PRICE
    57.40 €
    55.70 € / 1 L
    Save 1.70 €
    2% Off
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1310-73-2 | HNaO | 39.997 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Volumetric standard solution
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sodium hydroxide
Color Colorless
CAS 1310-73-2, 7732-18-5
Molecular Formula HNaO
Linear Formula NaOH
Concentration 0.998 to 1.002N (20°C)
Fieser 01,1083; 05,616; 07,336; 08,460
Density 1.0400g/mL
Merck Index 15, 8761
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Quantity 1 L
SMILES [OH-].[Na+]
IUPAC Name sodium;hydroxide
Molecular Weight (g/mol) 39.997
PubChem CID 14798
ChEBI CHEBI:32145
Formula Weight 40
Packaging Plastic Bottle
Physical Form Liquid
Show More Show Less
compliance-icons
Product Identifier
  • Sodium hydroxide, 1N standard solution
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Sodium hydroxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.