missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, purum p.a, >97%, Solstice
BIOPHARMA

Product Code. 15646830 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
1 kg
5 kg
25 kg
Packaging:
Poly bottle
Unit Size:
1kg
25kg
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15646830 5 kg Poly bottle 5kg
15626830 1 kg Poly bottle 1kg
15636830 25 kg Poly bottle 25kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15646830 Supplier Honeywell-Fluka Supplier No. 716945KG
  • ONLINE EXCLUSIVE PRICE
    148.00 €
    133.33 € / 5kg
    Save 14.67 €
    9% Off
 Restricted Item
Request Bulk or Custom Format
Currently Unavailable: We have no information when this product will be available.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Tested according to Ph.Eur.

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1310-73-2
Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
MDL Number MFCD00003548
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]

Specifications

pH Alkaline
Color Colorless
Boiling Point 1390°C
Melting Point 319°C
Linear Formula NaOH
UN Number UN1823
Density 2.13g/cm3
Quantity 5 kg
Formula Weight 40g/mol
Percent Purity >97%
Grade Reagent
Packaging Poly bottle
Vapor Density >1 (vs air)
Vapor Pressure 18mmHg (20°C), 3mmHg (37°C)
Physical Form Solid
Chemical Name or Material Sodium hydroxide
Show More Show Less
compliance-icons
Product Identifier
  • Sodium hydroxide
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Sodium hydroxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.