missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, purum p.a., ≥98% (T), ACS Reagent, Honeywell™

Product Code. 15696820 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
250 g
1 kg
5 kg
6 x 1 kg
Unit Size:
1kg
250g
5kg
Pack of 6
This item is not returnable. View return policy

Product Code. 15696820

Brand: Honeywell Research Chemicals 716916X1KG

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Purum p.a.,≥98.0% (T), beads (small)

  • Products with a guarantee certificate and/or suitable for the stated analytical application.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1310-73-2
Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
MDL Number MFCD00003548
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]

Specifications

Linear Formula NaOH
UN Number UN1823
Quantity 6 x 1 kg
Formula Weight 40
Percent Purity ≥98.0% (T)
Vapor Density >1 (vs air)
Vapor Pressure 3 mmHg (37°C), 18 mmHg (20°C)
Chemical Name or Material Sodium hydroxide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.