missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Comprehensive portfolio for the electronics and semiconductor industry
- For scientists engaged in education, research, development, prototyping and small scale fabrication of high tech micro- and nanochips
- Superior purity, stability and exceptional lot-to-lot consistency
- Includes high purity products required for high performance applications such as etchants, solvents or hexamethyldisilazane (HMDS)
Specifications
Specifications
| Linear Formula | NaOH |
| UN Number | 1823 |
| Solubility Information | Soluble in water, 1260 g/L at 20°C |
| Quantity | 500 g |
| Formula Weight | 40g/mol |
| Percent Purity | 99.99% (trace metals basis) |
| Grade | Semiconductor |
| Vapor Density | >1 (vs air) |
| Vapor Pressure | 18mmHg (20°C), 3mmHg (37°C) |
| Chemical Name or Material | Sodium Hydroxide |
Safety and Handling
Product Identifier
- Sodium hydroxide
- Danger
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : Sodium hydroxide
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?