missing translation for 'onlineSavingsMsg'
Learn More

Sodium hydroxide solution, 0.05M, Honeywell Fluka™

Product Code. 15634880 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
500 mL
1 L
6 x 1 L
Unit Size:
1 L
500mL
Pack of 6
This item is not returnable. View return policy

Product Code. 15634880

Brand: Honeywell-Fluka 352496X1L

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Volumetric, 0.05 M NaOH (0.05N)

Honeywell´s high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use

 

  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations

 


 

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1310-73-2
Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
MDL Number MFCD00003548
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]

Specifications

Linear Formula NaOH
UN Number NONH for all modes of transport
Quantity 6 x 1 L
Formula Weight 40
Vapor Pressure 3 mmHg (37°C)
Chemical Name or Material Sodium hydroxide solution, volumetric, 0.05 M NaOH (0.05N)
compliance-icons
Product Identifier
  • Sodium hydroxide solution
Signal Word
  • Warning
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
Hazard Statement
  • H290-May be corrosive to metals.
Precautionary Statement
  • P234-Keep only in original container.
Supplemental information
  • MIXTURE LIST-Contains : sodium hydroxide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.