missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide Solution, 0.111M (0.111N), for analysis of milk, Fisher Chemical™

Product Code. 15968776 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 L
5 L
Unit Size:
1 L
5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15968776 5 L 5 L
15958776 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15968776 Supplier Fisher Chemical Supplier No. J/7670/21
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Volumetric sodium hydroxide solution suitable for the determination of the acidity of milk.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 5 L
Normality 0.111
Packaging HDPE Plastic Bottle
Traceability to NIST Traceable to NIST, used to titrate the lactic acid present in milk
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.