missing translation for 'onlineSavingsMsg'
Learn More

Sodium hydroxide solution, Volumetric, Reag. Ph. Eur., 0.1 M NaOH (0.1N), Honeywell Fluka™

Product Code. 15654910 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
1 L
5 L
10 L
4 x 5 L
6 x 1 L
Packaging:
Plastic Bottle
VOLPAC™ CONTAINER
Unit Size:
1 L
10 L
5 L
Pack of 4
Pack of 6
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15654910 5 L Plastic Bottle 5 L
15634910 1 L Plastic Bottle 1 L
15664910 5 L VOLPAC™ CONTAINER 5 L
15674910 6 x 1 L Plastic Bottle Pack of 6
15624910 10 L VOLPAC™ CONTAINER 10 L
15644910 4 x 5 L - Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 15654910 Supplier Honeywell-Fluka Supplier No. 352635L
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Volumetric, Reag. Ph. Eur., 0.1 M NaOH (0.1N)

Honeywell´s high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use

 

  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations

 


 

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1310-73-2
Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
MDL Number MFCD00003548
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]

Specifications

Linear Formula NaOH
Concentration 0.1M
UN Number NONH for all modes of transport
Quantity 5 L
Formula Weight 40g/mol
Packaging Plastic Bottle
Vapor Pressure 3 mmHg (37°C)
Chemical Name or Material Sodium hydroxide solution
compliance-icons
Product Identifier
  • Sodium hydroxide solution
Signal Word
  • Warning
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
Hazard Statement
  • H290-May be corrosive to metals.
Precautionary Statement
  • P234-Keep only in original container.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P406-Store in corrosive resistant/ container with a resistant inner liner.
Supplemental information
  • MIXTURE LIST-Contain: sodium hydroxide; caustic soda
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.