missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide Solution, 0.1M, Honeywell Fluka™

Product Code. 15613580 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2 L
2.5 L
6 x 500 mL
6 x 1 L
Unit Size:
1 L
2 L
2.5 L
500mL
Pack of 6
Product Code. Quantity unitSize
15613580 6 x 500 mL Pack of 6
15693570 500 mL 500mL
15663570 1 L 1 L
15683570 2 L 2 L
15673570 2.5 L 2.5 L
15603580 6 x 1 L Pack of 6
6 options
This item is not returnable. View return policy

Product Code. 15613580

Brand: Honeywell-Fluka 3194816X500ML

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Volumetric, 0.1 M NaOH (0.1N)

Honeywell´s high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use

 

  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations

 


 

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1310-73-2
Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
MDL Number MFCD00003548
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]

Specifications

Linear Formula NaOH
Concentration 0.1M
UN Number NONH for all modes of transport
Density 1g/mL at 25°C
Quantity 6 x 500 mL
Formula Weight 40g/mol
Vapor Density >1 (vs air)
Vapor Pressure 3mmHg (37°C)
Chemical Name or Material Sodium Hydroxide Solution
compliance-icons
Product Identifier
  • Sodium hydroxide solution
Signal Word
  • Warning
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
Hazard Statement
  • H290-May be corrosive to metals.
Precautionary Statement
  • P234-Keep only in original container.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P406-Store in corrosive resistant/ container with a resistant inner liner.
Supplemental information
  • MIXTURE LIST-Contains : sodium hydroxide; caustic soda
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.