missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide Solution 0.2M (0.2N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Fisher Chemical™

Product Code. 10040200 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
5 L
10 L
Packaging:
Bag in box
HDPE plastic bottle
Plastic container
Unit Size:
1 L
10 L
2.5 L
5 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10040200 2.5 L HDPE plastic bottle 2.5 L
10316190 1 L HDPE plastic bottle 1 L
10627032 5 L Plastic container 5 L
10131860 10 L Bag in box 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10040200 Supplier Fisher Chemical Supplier No. J/7640/17
 Restricted Item
Estimated Shipment: 17-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

HNaO, CAS Number-1310-73-2, sodium hydroxide, sodium hydroxide solution, sodium hydroxide na oh, sodium hydrate, soda, caustic, white caustic, soda lye, aetznatron, caustic soda, ascarite, 1L, CHEBI:32145, Colorless, 39.99g/mol, HEMHJVSKTPXQMS-UHFFFAOYSA-M, sodium;hydroxide, 3548, 39.997

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1310-73-2
Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
MDL Number 3548
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]

Specifications

pH 13
Color Colorless
Quantity 2.5 L
Formula Weight 39.99g/mol
Packaging HDPE plastic bottle
Physical Form Liquid
compliance-icons
Product Identifier
  • Sodium hydroxide, 0.2 N standard solution
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.