missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide Solution, 40% w/w (d=1.43), for the determination of nitrogen, Fisher Chemical™

Product Code. 15978776 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
5 L
Unit Size:
5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15978776 5 L 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15978776 Supplier Fisher Chemical Supplier No. S/4960/21
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Sodium hydroxide solution suitable for use as a titrant for the determination of nitrogen by Kjeldahl method.

TRUSTED_SUSTAINABILITY

Specifications

Assay Percent Range 40% (w/w)
Density 1.43
Quantity 5 L
Packaging HDPE Plastic Bottle
Physical Form Liquid
compliance-icons
Product Identifier
  • Sodium hydroxide, 40% w/w
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.