missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium hydroxide solution 50% (w/w), BAKER ANALYZED™, J.T. Baker™
Click to view available options
1 L
Description
- IMPORTANT: Material will freeze if stored below 60°F (16°C). Protect from air to avoid absorption of carbon dioxide.
Specifications
Boiling Point | 119°C to 135°C (1013 hPa) |
CAS | 1310-73-2 |
Melting Point | 9°C to 10°C |
Assay Percent Range | 49.5 to 50.5% |
Molecular Formula | HNaO |
MDL Number | MFCD00003548 |
UN Number | 1824 |
Density | 1.53 g/cm³ (20°C) |
Synonym | Caustic soda |
InChI Key | HEMHJVSKTPXQMS-UHFFFAOYSA-M |
Show More |
Safety and Handling
Product Identifier
- 1LT SODIUM HYDROXIDE 50%
- Danger
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : sodium hydroxide
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'