missing translation for 'onlineSavingsMsg'
Learn More

Sodium hypochlorite solution, Reagent grade, Available chlorine 10 to 15%, Solstice

Product Code. 15685490
Change view
Click to view available options
Quantity:
250 mL
1 L
5 L
Unit Size:
1 L
250mL
5 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15685490 250 mL 250mL
15675490 1 L 1 L
15695490 5 L 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15685490 Supplier Fluka Supplier No. 425044250ML
 Restricted Item
Estimated Shipment: 24-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Reagent Grade, available chlorine 10-15 %

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7681-52-9
Molecular Formula ClNaO
Molecular Weight (g/mol) 74.439
MDL Number MFCD00011120
InChI Key SUKJFIGYRHOWBL-UHFFFAOYSA-N
Synonym sodium hypochlorite, antiformin, sodium oxychloride, chlorox, clorox, javex, javelle water, hypochlorous acid, sodium salt, hypochlorite sodium, carrel-dakin solution
PubChem CID 23665760
ChEBI CHEBI:32146
IUPAC Name sodium;hypochlorite
SMILES [O-]Cl.[Na+]

Specifications

Physical Form Liquid
Quantity 250 mL
Linear Formula NaClO
UN Number UN1791
Formula Weight 74.44g/mol
Percent Purity 32% to 35%
Grade Reagent
Concentration or Composition (by Analyte or Components) Available chlorine 10 to 15%
Density 1.206g/mL at 25°C
Chemical Name or Material Sodium hypochlorite solution
compliance-icons compliance-icons
Product Identifier
  • Sodium hypochlorite solution
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H410-Very toxic to aquatic life with long lasting effects.
  • EUH031-Contact with acids liberates toxic gas.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: sodium hypochlorite,solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.